Orders LinkContactUsLinkComponent
Pasteur pipettes
Pasteur pipettes
Pasteur pipettes
Supplier:  DELTALAB
Certificates
Pasteur pipettes
Pasteur pipettes
2 Options Available Below
Product Details & Documents
PE, transparent.
  • Made of inert polyethylene
  • Sterile models are sterilised by ethylene oxide
  • Volumetric tolerance: ±10%
Documents
Product Family Options
Product Information
  • Capacity
    Drop vol.
    Bulb draw
    Stem Ø
    Length
    Sterile
    Packed
    Availability
    Price
  • Fine tip pipette
    1,5 ml
    20 µl
    1 ml
    3 mm
    104 mm
    +
    Individually wrapped
    Log in for availability
    Log in for availability
    1 * 500 items
    1 * 500 items - 327,00 €
    327,00 €
    1 * 500 items
       327,00 €
  • DELT200005
    Pasteur pipette, general purpose
    Pasteur pipette, general purpose
    4 ml
    40 µl
    3,1 ml
    2,5 mm
    150 mm
    -
    6×500
    Discontinued
  • Fine tip pipette
    5,5 ml
    20 µl
    3 ml
    3 mm
    153 mm
    +
    Individually wrapped
    Log in for availability
    Log in for availability
    1 * 500 items
    1 * 500 items - 353,00 €
    353,00 €
    1 * 500 items
       353,00 €