Orders LinkContactUsLinkComponent
Buckets, Round
 :   DELTALAB
Buckets, Round
 :  DELTALAB
Buckets, Round
HDPE, white.
  • Good chemical and temperature resistance
  • Moulded-in graduations
  • With spout
For industrial and laboratory applications. With red stainless metal handle. Very sturdy construction.
Product Family Options
  • Colour
    Capacity
    Ø ext.
    Height
  • White
    9 L
    270 mm
    250 mm
    1 * 1 items
    1 * 1 items - 24,90 €
    24,90 €
    1 * 1 items
       24,90 €
  • White
    12 L
    300 mm
    280 mm
    1 * 1 items
    1 * 1 items - 27,60 €
    27,60 €
    1 * 1 items
       27,60 €
  • White
    17 L
    340 mm
    310 mm
    1 * 1 items
    1 * 1 items - 33,70 €
    33,70 €
    1 * 1 items
       33,70 €