Finger bandages, Aluderm®-aluplast
SOEH1009163
: SOEHNGEN
- Description:Finger bandages
- L×W:120×20 mm
- Pk:10
These bandages with aluderm® compress protect the wound. The sufficient length of the adhesive fabric means that the bandage can be wound around the finger several times, giving the wound compress a safe and fixed hold without impeding the finger function.