PCR microtubes and strips
8-strip PCR tubes, without caps, Clear, 0,2 ml, 125 tubes/pack
Catalog # 732-2463
Supplier: BIOTIX
Jump to:
Product Details & Documents
Medical grade virgin polypropylene.
- Excellent chemical and temperature resistance
- Biologically inert
- BioReady™ certified free from DNase, RNase and endotoxin
Superior clarity for sample visualisation.
Specifications
- Packed:125 tubes/pack
- Capacity:0,2 ml
- Colour:Clear
- Description:8-strip PCR tubes, without caps