Orders LinkContactUsLinkComponent
Racks for cryogenic vials, polycarbonate
Supplier:   DELTALAB
Racks for cryogenic vials, polycarbonate
Supplier:  DELTALAB
Certificates
Racks for cryogenic vials, polycarbonate
4 Options Available Below
Product Details & Documents
These cryovials storage boxes are made of extra strong polycarbonate, designed to be used at temperatures between −190 and 121 °C. Transparent covers are printed with a series of squares and are numbered from 1 to 81 for better inventory control.
  • Can be immersed in liquid nitrogen
  • Stackable and have air vents
  • Covers are keyed to the base to prevent misalignment
Boxes made to accept 81 tubes have a white base and a coloured grid (in which each well is numbered).
 : Each box is supplied with an innovative vial picker for easy vial removal.
Product Family Options
Product Information
  • For tubes
    No. of Holes
    Colour
    W×D×H
    Availability
    Price
  • 1,2/2,0 ml
    81
    Yellow
    133×133×52 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 61,00 €
    61,00 €
    1 * 1 items
       61,00 €
  • 1,2/2,0 ml
    81
    Red
    133×133×52 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 61,00 €
    61,00 €
    1 * 1 items
       61,00 €
  • 1,2/2,0 ml
    81
    Green
    133×133×52 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 59,00 €
    59,00 €
    1 * 1 items
       59,00 €
  • 3,4/5,0 ml
    81
    Yellow
    133×133×95 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 63,20 €
    63,20 €
    1 * 1 items
       63,20 €