Orders LinkContactUsLinkComponent
Tube racks, universal
Tube racks, universal
Tube racks, universal
Supplier:  DELTALAB
Certificates
Tube racks, universal
Tube racks, universal
3 Options Available Below
Product Details & Documents
PP, linkable to other racks.
  • Versatile rack with different capacities on each of the four sides
  • Alpha-numeric matrix on the four sides for better sample location
  • Suitable for freezers and autoclaves
Documents
Product Family Options
Product Information
  • Colour
    No. of Holes
    For tubes
    Material
    W×D×H
    Availability
    Price
  • Natural
    32/32/12/4
    0,5/1,5-2,0/15/50 ml
    PP
    172×50×90 mm
    Log in for availability
    Log in for availability
    1 * 5 items
    1 * 5 items - 186,00 €
    186,00 €
    1 * 5 items
       186,00 €
  • Pink
    32/32/12/4
    0,5/1,5-2,0/15/50 ml
    PP
    172×50×90 mm
    Log in for availability
    Log in for availability
    1 * 5 items
    1 * 5 items - 186,00 €
    186,00 €
    1 * 5 items
       186,00 €
  • Yellow
    32/32/12/4
    0,5/1,5-2,0/15/50 ml
    PP
    172×50×90 mm
    Log in for availability
    Log in for availability
    1 * 5 items
    1 * 5 items - 186,00 €
    186,00 €
    1 * 5 items
       186,00 €