ContactUsLinkComponent
Examination gloves, PVC, Unigloves® Premium
Examination gloves, PVC, Unigloves® Premium
Certificates
undefined
Examination gloves, PVC, Unigloves® Premium
Specifications
  • Catalog No:
  • UNIG1922
  • UNIG1923
  • UNIG1924
  • UNIG1925
  • Ambidextrous / hand-specific:
  • Ambidextrous
  • Ambidextrous
  • Ambidextrous
  • Ambidextrous
  • Disposable / reusable:
  • Disposable
  • Disposable
  • Disposable
  • Disposable
  • Latex / latex-free:
  • Latex-free
  • Latex-free
  • Latex-free
  • Latex-free
  • Powdered / powder-free:
  • Powder-free
  • Powder-free
  • Powder-free
  • Powder-free
  • Finish:
  • Smooth
  • Smooth
  • Smooth
  • Smooth
  • Cuff style:
  • Beaded
  • Beaded
  • Beaded
  • Beaded
  • Cuff thickness:
  • 0,08 - 0,14 mm
  • 0,08 - 0,14 mm
  • 0,08 - 0,14 mm
  • 0,08 - 0,14 mm
  • Finger thickness:
  • 0,08 - 0,13 mm
  • 0,08 - 0,13 mm
  • 0,08 - 0,13 mm
  • 0,08 - 0,13 mm
  • Palm thickness:
  • 0,08 - 0,14 mm
  • 0,08 - 0,14 mm
  • 0,08 - 0,14 mm
  • 0,08 - 0,14 mm
  • EN norm:
  • EN 455
  • EN 455
  • EN 455
  • EN 455
  • Food contact approved:
  • +
  • +
  • +
  • +