Orders LinkContactUsLinkComponent
Scoops
Scoops
Supplier:   LAMAPLAST
Scoops
Supplier:  LAMAPLAST
Certificates
Scoops
4 Options Available Below
Product Details & Documents
These scoops are made of high density PE. Ideal for use in food applications.
  • For withdrawal of solid and granulates samples
  • Rounded bottom
Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904
Documents
Product Family Options
Product Information
  • Capacity
    Length
    Material
    Colour
    Availability
    Price
  • 90 ml
    180 mm
    HDPE
    White
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 4,80 €
    4,80 €
    1 * 1 items
       4,80 €
  • 250 ml
    240 mm
    HDPE
    White
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 5,30 €
    5,30 €
    1 * 1 items
       5,30 €
  • 600 ml
    300 mm
    HDPE
    White
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 6,10 €
    6,10 €
    1 * 1 items
       6,10 €
  • 1100 ml
    360 mm
    HDPE
    White
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 8,20 €
    8,20 €
    1 * 1 items
       8,20 €