ContactUsLinkComponent
Atlas and axis
Atlas and axis
Catalog # BINH1000141
Supplier:  3B Scientific
undefined
Atlas and axis
Catalog # BINH1000141
Supplier:  3B Scientific
Specifications
  • Weight:
    100 g
  • Description:
    Atlas and axis
  • Pk:
    1
Specifications
About this item
This realistic replica is a great way to study the anatomy of the human atlas and axis bones.
Bones are wire mounted together on a removable stand.