ContactUsLinkComponent
Classic flexible spine model with femur heads
Classic flexible spine model with femur heads
Catalog # BINH1000122
Supplier:  3B Scientific
undefined
Classic flexible spine model with femur heads
Catalog # BINH1000122
Supplier:  3B Scientific
Specifications
  • Weight:
    2,1 kg
  • Description:
    Spine model with femur heads (83 cm)
  • Pk:
    1
Specifications
About this item
The spine contains these features:
  • Full pelvis and occipital plate
  • Fully flexible mounting in spine
  • L3-L4 disc prolapsed on spine
  • Spinal nerve exits
  • Cervical vertebral artery
  • Male pelvis
  • Movable mounted femur heads
The spine for patient education is the most affordable spine model. This spine model is fully flexible and designed for hands-on demonstrations.
Weight: 2,1 kg
Ordering information: Stand is not included.