J.T.Baker® BAKERBOND spe™ C18, SPE Columns
: Avantor
BAKERBOND spe™ columns are disposable PP or glass columns pre-packed with high capacity sorbents contained between two 20 µm PE frits.
- Silica based
- Extraction of compounds from sample solution volumes ranging from a few hundred µl up to 1 L
- Processing by vacuum or positive pressure
General applications: Non ionic, non polar to moderately polar analytes.
These SPE columns can be used in reversed phase mode.
: Certain columns are available in a foil-lined fibre drum called the Jumbo Pack. The columns are loose packed in the foil liner to eliminate the inconvenience of opening individual pouches.
- Catalog No:
- 7334-07
- 7334-06
- 7020-02
- 7020-06
- 7020-01
- 7020-00
- 7334-04
- 7020-13
- 7334-08
- 7334-03
- 7020-23
- 7020-03
- 7020-08
- 7020-27
- 7020-26
- 7020-07
- Carbon load:
- 17,2%
- 17,2%
- 17,2%
- 17,2%
- 17,2%
- 17,2%
- 17,2%
- 17,2%
- 17,2%
- 17,2%
- 17,2%
- 17,2%
- 17,2%
- 17,2%
- 17,2%
- 17,2%
- Endcapped:
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
Frequently Bought Together