Orders LinkContactUsLinkComponent
QuickTask™ flat mops
QuickTask™ flat mops
 :   CONTEC
QuickTask™ flat mops
 :  CONTEC
QuickTask™ flat mops
QuickTask™ flat mops
QuickTask™ mopping system. The QuickTask™ product line consists of a variety of knitted polyester, polyester/cellulose, microfibre/foam and microfibre flat mop heads that are easily installed onto a hinged mop head frame and QuickConnect interchangeable handle. QuickTask™ mop head frames are available in stainless steel or durable, autoclavable polypropylene.
  • All mop head covers are compatible with a wide range of chemicals and disinfectants
  • Ideally suited for cleaning floors, including perforated cleanroom floors and grates
  • TASK0300 is ideal for effective cleaning of polymeric flooring
QuickTask™ mop covers:
Polyester knit mop covers TASK0200 and TASK0250: These economical disposable mop covers are the lightest in the QuickTask™ range. Made of sorbent multi layer knitted polyester that is optimised for use on smooth surfaces and applies solutions evenly while removing contaminants. Supplied cleanroom laundered for more critical environments. Designed for single-use.
Looped Microfibre mop cover TASK0300: The most sorbent QuickTask™ mop cover, the small loop microfibre construction of TASK0300™ provides exceptional performance in removing liquids, soiling, residues and other contaminants from most surfaces. Microfibre construction for exceptional sorbency and mop-to-dry performance on wet surfaces. Can be laundered and autoclaved. Especially effective on polymeric flooring.
Looped polyester knit mop heads TASK0400: The cleanest and most durable mop head in the range, the medium length loops will conform to almost any shape, making it especially suitable for coving bases and wall/floor joints. Laundered and packed in an ISO Class 4 Cleanroom for a very low particulate level. Knitted tube microfibre polyester construction provides exceptional sorbency and drying of wet surfaces. Medium-loop design allows easy contact with irregular surfaces. Can be repeatedly laundered and autoclaved.
Polyester/cellulose blend mop cover TASK0500 and TASK0550: The lightest mop cover in the QuickTask™ range, TASK0500 and TASK0550 feature a polyester/cellulose cover over a polyester fibrefill core. Ideal for cost effective single-use. Highly sorbent mop cover is ideal for removing light soiling, residues and biofilms.
Polyester/cellulose blend mop cover TASK0600 and TASK0650: The unique microfibre face with foam core construction ensures excellent floor contact, sorbency and superior surface cleaning. Ribbed microfibre allows for exceptional cleaning, solution application and mop-to-dry performance. TASK0650 is validated sterile. Durable and economical, suitable for critical environments.
QuickTask™ mop head frames and QuickConnect™ handles:
Stainless steel and autoclavable polypropylene handles for QuickTask™ mops. Durable and lightweight hinged frames in stainless steel or blue polypropylene co-polymer. Quickly and easily slip into QuickTask™ mop head covers. Compatible with all common disinfectant solutions and most common solvents. Mop head frame 2749 is compatible with all Contec QuickConnect™ handles. All QuickTask™ hardware are fully autoclavable.
Looped polyester knit mop heads TASK0400: The cleanest and most durable mop head in the range, the medium length loops will conform to almost any shape, making it especially suitable for coving bases and wall/floor joints. Laundered and packed in an ISO Class 4 Cleanroom for a very low level of particulates. Knitted tube microfiber polyester construction provides exceptional sorbency and drying of wet surfaces. Medium-loop design allows easy contact with irregular surfaces. Can be repeatedly laundered and autoclaved.
Product Family Options
QuickTask™ mops
  • Description
    Colour
    Dimensions
    L×W
    Length
    Packed
  • QuickTask™ flat mop cover with pockets, non sterile, TASK0200
    White
    410×150 mm
    48 unit/case
    1 * 48 items
    1 * 48 items - 419,00 €
    419,00 €
    1 * 48 items
       419,00 €
  • QuickTask™ flat mop cover with pockets, sterile (gamma irradiated), TASK0250
    White
    410×150 mm
    40 unit/case
    1 * 40 items
    1 * 40 items - 476,00 €
    476,00 €
    1 * 40 items
       476,00 €
  • QuickTask™ looped microfibre mop head, autoclavable and non sterile, TASK0300
    Blue
    410×150 mm
    410×150 mm
    40 unit/case
    1 * 40 items
    1 * 40 items - 538,00 €
    538,00 €
    1 * 40 items
       538,00 €
  • QuickTask™ flat mop cover with pockets, non sterile, TASK0400
    White
    410×150 mm
    50 unit/case
    1 * 50 items
    1 * 50 items - 1 140,00 €
    1 140,00 €
    1 * 50 items
       1 140,00 €
  • QuickTask™ polyester/cellulose flat mop head, non sterile, TASK0500
    White
    410×150 mm
    410×150 mm
    48 unit/case
    1 * 48 items
    1 * 48 items - 247,00 €
    247,00 €
    1 * 48 items
       247,00 €
  • QuickTask™ polyester/cellulose flat mop head, sterile (gamma irradiated), TASK0550
    White
    410×150 mm
    40 unit/case
    1 * 40 items
    1 * 40 items - 318,00 €
    318,00 €
    1 * 40 items
       318,00 €
QuickTask™ frames and QuickConnect™ handles
  • Description
    Colour
    Dimensions
    L×W
    Length
    Packed
  • QuickTask™ magnetic mop frame, stainless steel
    Chrome
    394 mm
    1 * 1 items
    1 * 1 items - 375,00 €
    375,00 €
    1 * 1 items
       375,00 €
  • QuickTask™ magnetic mop frame, polyproylene
    Blue
    394 mm
    1 * 1 items
    1 * 1 items - 204,00 €
    204,00 €
    1 * 1 items
       204,00 €
  • QuickTask™ polypropylene flat hinged mop head frame 2740, attaches easily to stainless steel handle 2725E
    Blue
    400 mm
    1 * 1 items
    1 * 1 items - 75,60 €
    75,60 €
    1 * 1 items
       75,60 €
  • QuickConnect™, stainless steel handle, lightweight, hanging loop for QuickTask™ mop head frame
    Grey
    406 mm
    1 * 1 items
    1 * 1 items - 34,60 €
    34,60 €
    1 * 1 items
       34,60 €
  • QuickConnect™ anodised aluminium telescopic handle
    Grey
    406 - 762 mm
    1 * 1 items
    1 * 1 items - 175,00 €
    175,00 €
    1 * 1 items
       175,00 €
  • QuickConnect™ anodised aluminium telescopic handle
    Grey
    1270 - 2337 mm
    1 * 1 items
    1 * 1 items - 210,00 €
    210,00 €
    1 * 1 items
       210,00 €
  • QuickConnect™ stainless steel telescopic handle
    Grey
    1270 - 2337 mm
    1 * 1 items
    1 * 1 items - 497,00 €
    497,00 €
    1 * 1 items
       497,00 €
  • QuickConnect™ stainless steel handle, for QuickTask™ polypropylene mop head frame 2740
    Grey
    1520 mm
    1 * 1 items
    1 * 1 items - 108,00 €
    108,00 €
    1 * 1 items
       108,00 €