Orders LinkContactUsLinkComponent
Slide boxes
Slide boxes
Slide boxes
Supplier:  DELTALAB
Certificates
Slide boxes
2 Options Available Below
Product Details & Documents
Stackable items to gain space on the bench.
  • Translucent lid to see if the box is empty or not
  • No numbering of the slots
Documents
Product Family Options
Product Information
  • Colour
    Material
    No. of slides
    W×D×H
    Availability
    Price
  • White
    PS
    25
    105×88×32 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 16,80 €
    16,80 €
    1 * 1 items
       16,80 €
  • White
    PS
    50
    200×88×32 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 23,80 €
    23,80 €
    1 * 1 items
       23,80 €