Reagent reservoirs, disposable
Catalog # VIST3054-1004
Supplier: VISTALAB TECHNOLOGIES
Specifications
- Packed:Bulk
- Sterile:-
- Type:Divided
- Capacity:25 ml
- Seq:6
- Pk:100
Specifications
About this item
These reservoirs made of PS feature a trough-within-a-trough design to provide the lowest residual volume in the industry, meaning maximal reagent recovery.
- Pour-off spouts at every corner
- Wider base for stability
- Free of animal products
- Sterile reservoirs are certified RNase/DNase free and non pyrogenic