Vinometer according to Bernadotte
HECH44200010
- Description:Vinometer
- Material:Glass
- Model:Bernadotte
- Pk:1
- Range:0 − 25
This instrument is designed for the accurate testing the percentage of alcohol in wine.
This instrument is designed for the accurate testing the percentage of alcohol in wine.