Orders LinkContactUsLinkComponent
Hydrometers
Hydrometers
Supplier:   Amarell
Hydrometers
Supplier:  Amarell
Certificates
Hydrometers
3 Options Available Below
Product Details & Documents
Hydrometers for special applications.
Product Family Options
Product Information
  • Range
    Division
    Length
    Availability
    Price
  • AMARH860616
    Hydrometer for brine acc. to Bischoff
    Hydrometer for brine acc. to Bischoff
    0 - 27 %
    1 %
    270 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 38,90 €
    38,90 €
    1 * 1 items
       38,90 €
  • AMARH860660
    Hydrometer for potassium hydroxide (KOH)
    Hydrometer for potassium hydroxide (KOH)
    0 - 50 %
    1 %
    280 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 34,90 €
    34,90 €
    1 * 1 items
       34,90 €
  • AMARH860640
    Hydrometer for phosphoric acid (H₃PO₃)
    Hydrometer for phosphoric acid (H₃PO₃)
    0 - 75 %
    1 %
    300 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 44,00 €
    44,00 €
    1 * 1 items
       44,00 €