ContactUsLinkComponent
Slide mailer, Corning® Gosselin™
Slide mailer, Corning® Gosselin™
Catalog # GOSSPLP-01
Supplier:  Corning
   Low Manufacturing Impact
undefined
Slide mailer, Corning® Gosselin™
Catalog # GOSSPLP-01
Supplier:  Corning
   Low Manufacturing Impact
Specifications
  • No. of slides:
    2
  • Colour:
    Natural
  • Description:
    Slide mailers
  • Material:
    PP
  • W×D×H:
    85×55×5 mm
  • Environmentally Preferable:
    Low Manufacturing Impact
  • Environmental certification:
    RECs, ISO 14001 - Borre
  • Pk:
    700
Specifications
About this item
This slide mailer is suitable for applications such as clinical, dairy, environment, industries and microbiology.