Orders LinkContactUsLinkComponent
Small reaction vials, Reacti-Vial™
Small reaction vials, Reacti-Vial™
Certificates
Small reaction vials, Reacti-Vial™
Small reaction vials, Reacti-Vial™
8 Options Available Below
Product Details & Documents
These small reaction vials have an internal cone designed to make small-sample collection and handling easy and convenient. The cone feature is particularly useful for removing small quantities of sample with a syringe.
  • Extra thick glass wall magnifies the sample ideal for observing chemical reactions
  • Amber vials are light-sensitive to well protect the compounds
These vials are ideal for residue isolation, derivative preparation, maximum sample retrieval, moisture protection, sample storage, purification, precipitations, centrifugations and solvent separations.
 : Vials are supplied complete with open-top screw caps and teflon/rubber laminated discs.
Product Family Options
Amber
Product Information
  • Capacity
    Ø×H
    Colour
    Cap included
    Availability
    Price
  • HYPETS-13097
    Vial, inside Ø 11 mm, thread style 425-13
    Vial, inside Ø 11 mm, thread style 425-13
    1,00 ml
    13×45 mm
    Amber
    Yes
    Log in for availability
    Log in for availability
    1 * 12 items
    1 * 12 items - 232,00 €
    232,00 €
    1 * 12 items
       232,00 €
  • HYPETS-13099
    Vial, inside Ø 18 mm, thread style 425-20
    Vial, inside Ø 18 mm, thread style 425-20
    5,00 ml
    20×60 mm
    Amber
    Yes
    Log in for availability
    Log in for availability
    1 * 12 items
    1 * 12 items - 348,00 €
    348,00 €
    1 * 12 items
       348,00 €
Clear
Product Information
  • Capacity
    Ø×H
    Colour
    Cap included
    Availability
    Price
  • HYPETS-13100
    Vial, inside Ø 8 mm, thread style 425-8
    Vial, inside Ø 8 mm, thread style 425-8
    0,10 ml
    12×32 mm
    Clear
    Yes
    Log in for availability
    Log in for availability
    1 * 12 items
    1 * 12 items - 186,00 €
    186,00 €
    1 * 12 items
       186,00 €
  • HYPETS-13220
    Vial, inside Ø 11 mm, thread style 425-13
    Vial, inside Ø 11 mm, thread style 425-13
    0,30 ml
    13×32 mm
    Clear
    Yes
    Log in for availability
    Log in for availability
    1 * 12 items
    1 * 12 items - 186,00 €
    186,00 €
    1 * 12 items
       186,00 €
  • HYPETS-13221
    Vial, inside Ø 11 mm, thread style 425-13
    Vial, inside Ø 11 mm, thread style 425-13
    1,00 ml
    13×45 mm
    Clear
    Yes
    Log in for availability
    Log in for availability
    1 * 12 items
    1 * 12 items - 226,00 €
    226,00 €
    1 * 12 items
       226,00 €
  • HYPETS-13222
    Vial, inside Ø 18 mm, thread style 425-20
    Vial, inside Ø 18 mm, thread style 425-20
    3,00 ml
    20×47 mm
    Clear
    Yes
    Log in for availability
    Log in for availability
    1 * 12 items
    1 * 12 items - 271,00 €
    271,00 €
    1 * 12 items
       271,00 €
  • HYPETS-13223
    Vial, inside Ø 18 mm, thread style 425-20
    Vial, inside Ø 18 mm, thread style 425-20
    5,00 ml
    20×60 mm
    Clear
    Yes
    Log in for availability
    Log in for availability
    1 * 12 items
    1 * 12 items - 248,00 €
    248,00 €
    1 * 12 items
       248,00 €
  • HYPETS-13225
    Vial, inside Ø 22 mm, thread style 425-24
    Vial, inside Ø 22 mm, thread style 425-24
    10,00 ml
    25×69 mm
    Clear
    Yes
    Log in for availability
    Log in for availability
    1 * 12 items
    1 * 12 items - 462,00 €
    462,00 €
    1 * 12 items
       462,00 €