Orders LinkContactUsLinkComponent
Rod for centering
Rod for centering
15 mm, 300 mm, Stainless steel 18/10
Catalog # BOCH17031
Supplier:   BOCHEM
Rod for centering
Catalog # BOCH17031
Supplier:  BOCHEM
Rod for centering
Rod for centering
15 mm, 300 mm, Stainless steel 18/10
Price
77,60 €
Log in for availability
1 * 1 items
Product Details & Documents
Stainless steel.
Specifications
  • Ø:
    15 mm
  • Length:
    300 mm
  • Material:
    Stainless steel 18/10