ContactUsLinkComponent
Silver nitrate 1.88 mol/l (1.88 N) in aqueous solution
Corrosive
Environment
Silver nitrate 1.88 mol/l (1.88 N) in aqueous solution
Catalog # HACH2349432
Supplier:  Hach
CAS Number:  
undefined
Corrosive
Environment
Silver nitrate 1.88 mol/l (1.88 N) in aqueous solution
Catalog # HACH2349432
Supplier:  Hach
CAS Number:  
Specifications
  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Seq:
    2440
Specifications
About this item
 :
 :169.87 g/mol
Storage Temperature:Ambient
EINECS:231-853-9
UN:3264
ADR:8,II
 :GHS05
 :GHS09
CAS Number:7761-88-8
Formula:AgNO₃
MDL Number:MFCD00003414