Orders LinkContactUsLinkComponent
Assortment Boxes, PP
Assortment Boxes, PP
Supplier:   SEMADENI
Assortment Boxes, PP
Supplier:  SEMADENI
Certificates
Assortment Boxes, PP
Assortment Boxes, PP
6 Options Available Below
Product Details & Documents
These assortment boxes, made of PP, have a transparent stable hinged lid with lock and tamper-evident seals.
  • Robust and strong design
  • All models can be stacked together
  • Lid is fixed on each tray so that the contents don't mix even if box is turned upside down
Product Family Options
Product Information
  • Colour
    W×D×H
    Availability
    Price
  • SEMA1770
    Assortment box, with hinged lid and 4×1, 2×1 and 1×1 compartments
    Assortment box, with hinged lid and 4×1, 2×1 and 1×1 compartments
    Anthracite
    175×140×30 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 21,70 €
    21,70 €
    1 * 1 items
       21,70 €
  • 216-1268
    Assortment box, with hinged lid and 3×4 compartments
    Assortment box, with hinged lid and 3×4 compartments
    Anthracite
    175×140×30 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 21,20 €
    21,20 €
    1 * 1 items
       21,20 €
  • 216-7331
    Assortment box, with hinged lid and 4×1, 2×1, 2×1 and 1×1 compartments
    Assortment box, with hinged lid and 4×1, 2×1, 2×1 and 1×1 compartments
    Anthracite
    240×195×40 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 32,90 €
    32,90 €
    1 * 1 items
       32,90 €
  • SEMA1773
    Assortment box, with hinged lid and 3×6 compartments
    Assortment box, with hinged lid and 3×6 compartments
    Anthracite
    240×195×40 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 32,90 €
    32,90 €
    1 * 1 items
       32,90 €
  • 216-7332
    Assortment box, with hinged lid and 8×1, 3×1, 2×1, 1×1 and 1×1 compartments
    Assortment box, with hinged lid and 8×1, 3×1, 2×1, 1×1 and 1×1 compartments
    Anthracite
    340×260×55 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 60,00 €
    60,00 €
    1 * 1 items
       60,00 €
  • 215-0289
    Assortment box, with hinged lid and 4×8 compartments
    Assortment box, with hinged lid and 4×8 compartments
    Anthracite
    340×260×55 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 60,00 €
    60,00 €
    1 * 1 items
       60,00 €