Orders LinkContactUsLinkComponent
Erlenmeyer flasks with screw cap
Erlenmeyer flasks with screw cap
Supplier:   KAVALIER GLASS
Erlenmeyer flasks with screw cap
Supplier:  KAVALIER GLASS
Certificates
Erlenmeyer flasks with screw cap
Erlenmeyer flasks with screw cap
2 Options Available Below
Product Details & Documents
Borosilicate glass 3,3, clear, with teflon lined blue PP screw cap.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
Volume graduations and labelling area. Cap temperature resistant up to 140 °C.
Product Family Options
Product Information
  • Capacity
    Height
    Ext.Ø
    Thread
    Availability
    Price
  • 100 ml
    100 mm
    64 mm
    GL 25
    Log in for availability
    Log in for availability
    1 * 10 items
    1 * 10 items - 636,00 €
    636,00 €
    1 * 10 items
       636,00 €
  • 250 ml
    140 mm
    85 mm
    GL 32
    Discontinued
  • 1000 ml
    215 mm
    131 mm
    GL 32
    Log in for availability
    Log in for availability
    1 * 10 items
    1 * 10 items - 670,00 €
    670,00 €
    1 * 10 items
       670,00 €