Orders LinkContactUsLinkComponent
Separating funnel, pear shaped
Separating funnel, pear shaped
1000 ml, 29/32 NS
Catalog # KAVA632426355940
Supplier:   KAVALIER GLASS
Separating funnel, pear shaped
Catalog # KAVA632426355940
Supplier:  KAVALIER GLASS
Separating funnel, pear shaped
Separating funnel, pear shaped
1000 ml, 29/32 NS
Price
349,00 €
Log in for availability
1 * 1 items
Product Details & Documents
Made of borosilicate glass.
  • With stopper
  • Stopcock with glass key
  • Very good chemical and temperature resistance
Specifications
  • Socket size:
    29/32 NS
  • Capacity:
    1000 ml