Orders LinkContactUsLinkComponent
Jars, with screw cap
Jars, with screw cap
Supplier:   SEMADENI
Jars, with screw cap
Supplier:  SEMADENI
Certificates
Jars, with screw cap
Jars, with screw cap
6 Options Available Below
Product Details & Documents
PP, white.
  • Good chemical resistance
  • Good temperature resistance
Ideal for collecting, storing and transporting samples.
Product Family Options
Product Information
  • Capacity
    Ext.Ø
    Height
    Colour
    Availability
    Price
  • 6 ml
    28 mm
    18 mm
    White
    Log in for availability
    Log in for availability
    1 * 100 items
    1 * 100 items - 28,10 €
    28,10 €
    1 * 100 items
       28,10 €
  • 25 ml
    34 mm
    40 mm
    White
    Log in for availability
    Log in for availability
    1 * 100 items
    1 * 100 items - 37,60 €
    37,60 €
    1 * 100 items
       37,60 €
  • 35 ml
    38 mm
    48 mm
    White
    Log in for availability
    Log in for availability
    1 * 100 items
    1 * 100 items - 46,10 €
    46,10 €
    1 * 100 items
       46,10 €
  • 60 ml
    45 mm
    55 mm
    White
    Log in for availability
    Log in for availability
    1 * 100 items
    1 * 100 items - 68,30 €
    68,30 €
    1 * 100 items
       68,30 €
  • 125 ml
    56 mm
    67 mm
    White
    Log in for availability
    Log in for availability
    1 * 100 items
    1 * 100 items - 84,20 €
    84,20 €
    1 * 100 items
       84,20 €
  • 500 ml
    95 mm
    91 mm
    White
    Log in for availability
    Log in for availability
    1 * 100 items
    1 * 100 items - 275,00 €
    275,00 €
    1 * 100 items
       275,00 €