Orders LinkContactUsLinkComponent
Height adjustable worktable
Height adjustable worktable
Supplier:   TRESTON
Height adjustable worktable
Supplier:  TRESTON
Certificates
Height adjustable worktable
Height adjustable worktable
3 Options Available Below
Product Details & Documents
These worktables are a suitable solution for industrial laboratories and research and development centers. They are designed to accommodate the necessary measurement equipment and other tools ergonomically at a safe distance for use. These laboratory benches fit also to education purposes. Modularity and accessories are individually adjustable according the task and human requirements.
  • Robust, elegant, and very flexible
  • Easy to adapt to individual needs, new equipment, and changes in the working environment
  • With durable table frame
  • Leveling glides
Ordering information: Table top sold separately; contact your Avantor representative for more information.
Product Family Options
Product Information
  • W×D×H
    Availability
    Price
  • SOVEC10041037
    Concept table (stepless height adjustment)
    Concept table (stepless height adjustment)
    1500× 750× 670/1120 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items
  • SOVEC10141037
    Concept hand crank table (height adjustment with hand crank)
    Concept hand crank table (height adjustment with hand crank)
    1500× 750× 675/1025 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items
  • SOVEC10341037
    Concept motor table, (motorised height adjustment) ; control panel with three memory settings
    Concept motor table, (motorised height adjustment) ; control panel with three memory settings
    1500×750×700/1100 mm