Multipurpose trays, SP Bel-Art
BELAF162000011
- Capacity:8,5 l
- Colour:Blue
- Description:Multipurpose tray, with handle
- W×D×H:324×267×108 mm
- Pk:1
Trays made of PP.
- Stackable
- Top rim adds rigidity and provides handholds for transport
- Rounded inside corners make cleaning easy
Ideal for holding instruments while autoclaving and cleaning glassware to reduce chance of damage.