Laboratory coats, polyester/cotton, 2852
Supplier: Portwest
About this item
Standard V-necked coats, made of Fortis Plus fabric consisting of 65% polyester and 35% cotton.
- Concealed stud front
- Back vent
- Breast pocket, two lower patch pockets
- Excellent dye retention and fully pre-shrunk
Weight: 245 g/m²
27 Options Available Below
Product Details & Documents
Standard V-necked coats, made of Fortis Plus fabric consisting of 65% polyester and 35% cotton.
- Concealed stud front
- Back vent
- Breast pocket, two lower patch pockets
- Excellent dye retention and fully pre-shrunk
Weight: 245 g/m²
Specifications for Product Family
Specifications
- Catalog No:
- 113-8565
- 113-6080
- 113-6078
- 113-6077
- 113-6076
- 113-6079
- 113-6081
- 113-6082
- 113-8116
- 113-8117
- 113-7978
- 113-7652
- 113-7653
- 113-7654
- 113-7655
- 113-7656
- 113-7657
- 113-8566
- 113-8567
- 113-8568
- 113-8569
- 113-8570
- 113-9112
- 113-9113
- 113-9114
- 113-9111
- 113-9110
- Garment application:
- General Purpose|Medical|Laboratory|Clinical|Industrial
- General Purpose|Medical|Laboratory|Clinical|Industrial
- General Purpose|Medical|Laboratory|Clinical|Industrial
- General Purpose|Medical|Laboratory|Clinical|Industrial
- General Purpose|Medical|Laboratory|Clinical|Industrial
- General Purpose|Medical|Laboratory|Clinical|Industrial
- General Purpose|Medical|Laboratory|Clinical|Industrial
- General Purpose|Medical|Laboratory|Clinical|Industrial
- General Purpose|Medical|Laboratory|Clinical|Industrial
- General Purpose|Medical|Laboratory|Clinical|Industrial
- General Purpose|Medical|Laboratory|Clinical|Industrial
- General Purpose|Medical|Laboratory|Clinical|Industrial
- General Purpose|Medical|Laboratory|Clinical|Industrial
- General Purpose|Medical|Laboratory|Clinical|Industrial
- General Purpose|Medical|Laboratory|Clinical|Industrial
- General Purpose|Medical|Laboratory|Clinical|Industrial
- General Purpose|Medical|Laboratory|Clinical|Industrial
- General Purpose|Medical|Laboratory|Clinical|Industrial
- General Purpose|Medical|Laboratory|Clinical|Industrial
- General Purpose|Medical|Laboratory|Clinical|Industrial
- General Purpose|Medical|Laboratory|Clinical|Industrial
- General Purpose|Medical|Laboratory|Clinical|Industrial
- General Purpose|Medical|Laboratory|Clinical|Industrial
- General Purpose|Medical|Laboratory|Clinical|Industrial
- General Purpose|Medical|Laboratory|Clinical|Industrial
- General Purpose|Medical|Laboratory|Clinical|Industrial
- General Purpose|Medical|Laboratory|Clinical|Industrial
- Gender:
- For men
- For men
- For men
- For men
- For men
- For men
- For men
- For men
- For men
- For men
- For men
- For men
- For men
- For men
- For men
- For men
- For men
- For men
- For men
- For men
- For men
- For men
- For men
- For men
- For men
- For men
- For men
- Sleeve or leg length:
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Closure type:
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Fabric:
- 65% polyester and 35% cotton
- 65% polyester and 35% cotton
- 65% polyester and 35% cotton
- 65% polyester and 35% cotton
- 65% polyester and 35% cotton
- 65% polyester and 35% cotton
- 65% polyester and 35% cotton
- 65% polyester and 35% cotton
- 65% polyester and 35% cotton
- 65% polyester and 35% cotton
- 65% polyester and 35% cotton
- 65% polyester and 35% cotton
- 65% polyester and 35% cotton
- 65% polyester and 35% cotton
- 65% polyester and 35% cotton
- 65% polyester and 35% cotton
- 65% polyester and 35% cotton
- 65% polyester and 35% cotton
- 65% polyester and 35% cotton
- 65% polyester and 35% cotton
- 65% polyester and 35% cotton
- 65% polyester and 35% cotton
- 65% polyester and 35% cotton
- 65% polyester and 35% cotton
- 65% polyester and 35% cotton
- 65% polyester and 35% cotton
- 65% polyester and 35% cotton
- Weight:
- 245 g/m²
- 245 g/m²
- 245 g/m²
- 245 g/m²
- 245 g/m²
- 245 g/m²
- 245 g/m²
- 245 g/m²
- 245 g/m²
- 245 g/m²
- 245 g/m²
- 245 g/m²
- 245 g/m²
- 245 g/m²
- 245 g/m²
- 245 g/m²
- 245 g/m²
- 245 g/m²
- 245 g/m²
- 245 g/m²
- 245 g/m²
- 245 g/m²
- 245 g/m²
- 245 g/m²
- 245 g/m²
- 245 g/m²
- 245 g/m²
Specifications
Product Family Options
Product Information
- ColourClothing SizeLengthAvailabilityPrice
- Specifications:
More
Specifications
Cat. No.113-8565Garment applicationGeneral Purpose|Medical|Laboratory|Clinical|IndustrialGenderFor menSleeve or leg lengthLongClosure typePress studs / snapsFabric65% polyester and 35% cottonWeight245 g/m² - Specifications:
More
Specifications
Cat. No.113-6080Garment applicationGeneral Purpose|Medical|Laboratory|Clinical|IndustrialGenderFor menSleeve or leg lengthLongClosure typePress studs / snapsFabric65% polyester and 35% cottonWeight245 g/m² - Specifications:
More
Specifications
Cat. No.113-6078Garment applicationGeneral Purpose|Medical|Laboratory|Clinical|IndustrialGenderFor menSleeve or leg lengthLongClosure typePress studs / snapsFabric65% polyester and 35% cottonWeight245 g/m² - Specifications:
More
Specifications
Cat. No.113-6077Garment applicationGeneral Purpose|Medical|Laboratory|Clinical|IndustrialGenderFor menSleeve or leg lengthLongClosure typePress studs / snapsFabric65% polyester and 35% cottonWeight245 g/m² - Specifications:
More
Specifications
Cat. No.113-6076Garment applicationGeneral Purpose|Medical|Laboratory|Clinical|IndustrialGenderFor menSleeve or leg lengthLongClosure typePress studs / snapsFabric65% polyester and 35% cottonWeight245 g/m² - Specifications:
More
Specifications
Cat. No.113-6079Garment applicationGeneral Purpose|Medical|Laboratory|Clinical|IndustrialGenderFor menSleeve or leg lengthLongClosure typePress studs / snapsFabric65% polyester and 35% cottonWeight245 g/m² - Specifications:
More
Specifications
Cat. No.113-6081Garment applicationGeneral Purpose|Medical|Laboratory|Clinical|IndustrialGenderFor menSleeve or leg lengthLongClosure typePress studs / snapsFabric65% polyester and 35% cottonWeight245 g/m² - Specifications:
More
Specifications
Cat. No.113-6082Garment applicationGeneral Purpose|Medical|Laboratory|Clinical|IndustrialGenderFor menSleeve or leg lengthLongClosure typePress studs / snapsFabric65% polyester and 35% cottonWeight245 g/m² - Specifications:
More
Specifications
Cat. No.113-8116Garment applicationGeneral Purpose|Medical|Laboratory|Clinical|IndustrialGenderFor menSleeve or leg lengthLongClosure typePress studs / snapsFabric65% polyester and 35% cottonWeight245 g/m² - Specifications:
More
Specifications
Cat. No.113-8117Garment applicationGeneral Purpose|Medical|Laboratory|Clinical|IndustrialGenderFor menSleeve or leg lengthLongClosure typePress studs / snapsFabric65% polyester and 35% cottonWeight245 g/m² - Specifications:
More
Specifications
Cat. No.113-7978Garment applicationGeneral Purpose|Medical|Laboratory|Clinical|IndustrialGenderFor menSleeve or leg lengthLongClosure typePress studs / snapsFabric65% polyester and 35% cottonWeight245 g/m² - Specifications:
More
Specifications
Cat. No.113-7652Garment applicationGeneral Purpose|Medical|Laboratory|Clinical|IndustrialGenderFor menSleeve or leg lengthLongClosure typePress studs / snapsFabric65% polyester and 35% cottonWeight245 g/m² - Specifications:
More
Specifications
Cat. No.113-7653Garment applicationGeneral Purpose|Medical|Laboratory|Clinical|IndustrialGenderFor menSleeve or leg lengthLongClosure typePress studs / snapsFabric65% polyester and 35% cottonWeight245 g/m² - Specifications:
More
Specifications
Cat. No.113-7654Garment applicationGeneral Purpose|Medical|Laboratory|Clinical|IndustrialGenderFor menSleeve or leg lengthLongClosure typePress studs / snapsFabric65% polyester and 35% cottonWeight245 g/m² - Specifications:
More
Specifications
Cat. No.113-7655Garment applicationGeneral Purpose|Medical|Laboratory|Clinical|IndustrialGenderFor menSleeve or leg lengthLongClosure typePress studs / snapsFabric65% polyester and 35% cottonWeight245 g/m² - Specifications:
More
Specifications
Cat. No.113-7656Garment applicationGeneral Purpose|Medical|Laboratory|Clinical|IndustrialGenderFor menSleeve or leg lengthLongClosure typePress studs / snapsFabric65% polyester and 35% cottonWeight245 g/m² - Specifications:
More
Specifications
Cat. No.113-7657Garment applicationGeneral Purpose|Medical|Laboratory|Clinical|IndustrialGenderFor menSleeve or leg lengthLongClosure typePress studs / snapsFabric65% polyester and 35% cottonWeight245 g/m² - Specifications:
More
Specifications
Cat. No.113-8566Garment applicationGeneral Purpose|Medical|Laboratory|Clinical|IndustrialGenderFor menSleeve or leg lengthLongClosure typePress studs / snapsFabric65% polyester and 35% cottonWeight245 g/m² - Specifications:
More
Specifications
Cat. No.113-8567Garment applicationGeneral Purpose|Medical|Laboratory|Clinical|IndustrialGenderFor menSleeve or leg lengthLongClosure typePress studs / snapsFabric65% polyester and 35% cottonWeight245 g/m² - Specifications:
More
Specifications
Cat. No.113-8568Garment applicationGeneral Purpose|Medical|Laboratory|Clinical|IndustrialGenderFor menSleeve or leg lengthLongClosure typePress studs / snapsFabric65% polyester and 35% cottonWeight245 g/m² - Specifications:
More
Specifications
Cat. No.113-8569Garment applicationGeneral Purpose|Medical|Laboratory|Clinical|IndustrialGenderFor menSleeve or leg lengthLongClosure typePress studs / snapsFabric65% polyester and 35% cottonWeight245 g/m² - Specifications:
More
Specifications
Cat. No.113-8570Garment applicationGeneral Purpose|Medical|Laboratory|Clinical|IndustrialGenderFor menSleeve or leg lengthLongClosure typePress studs / snapsFabric65% polyester and 35% cottonWeight245 g/m² - Specifications:
More
Specifications
Cat. No.113-9112Garment applicationGeneral Purpose|Medical|Laboratory|Clinical|IndustrialGenderFor menSleeve or leg lengthLongClosure typePress studs / snapsFabric65% polyester and 35% cottonWeight245 g/m² - Specifications:
More
Specifications
Cat. No.113-9113Garment applicationGeneral Purpose|Medical|Laboratory|Clinical|IndustrialGenderFor menSleeve or leg lengthLongClosure typePress studs / snapsFabric65% polyester and 35% cottonWeight245 g/m² - Specifications:
More
Specifications
Cat. No.113-9114Garment applicationGeneral Purpose|Medical|Laboratory|Clinical|IndustrialGenderFor menSleeve or leg lengthLongClosure typePress studs / snapsFabric65% polyester and 35% cottonWeight245 g/m² - Specifications:
More
Specifications
Cat. No.113-9111Garment applicationGeneral Purpose|Medical|Laboratory|Clinical|IndustrialGenderFor menSleeve or leg lengthLongClosure typePress studs / snapsFabric65% polyester and 35% cottonWeight245 g/m² - Specifications:
More
Specifications
Cat. No.113-9110Garment applicationGeneral Purpose|Medical|Laboratory|Clinical|IndustrialGenderFor menSleeve or leg lengthLongClosure typePress studs / snapsFabric65% polyester and 35% cottonWeight245 g/m²