Buffer, TRIS solution pH 7 (1 mol/L), Trizma®, Sigma-Aldrich®
: Merck
- M-Clarity quality level:MQ200
Frequently Bought Together
Acetone ≥99.8%, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
Ethanol 70% ≥70% (v/v), TechniSolv®
: VWR Chemicals
Acetone ≥99%, TECHNICAL
: VWR Chemicals
VWR® Ice Buckets, Rectangular, with Lid
: VWR Collection
Ethanol eurodenatured ≥96%, TechniSolv® (1:1:1)
: VWR Chemicals
2-Propanol ≥99.8%, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
Water, AnalaR NORMAPUR® ISO 3696 Grade 3 analytical reagent
: VWR Chemicals
Ethanol absolute ≥99.5%, TechniSolv®, pure
: VWR Chemicals
Guanylate Cyclase α subunit (soluble) Polyclonal Antibody 1 * 500 µl
: Cayman Chemical