ContactUsLinkComponent
VWR® Q Path® Storage, 8 Drawers
VWR® Q Path® Storage, 8 Drawers
Catalog # 631-1050
Supplier:  VWR Collection
undefined
VWR® Q Path® Storage, 8 Drawers
Catalog # 631-1050
Supplier:  VWR Collection
Specifications
  • Description:
    Organiser with 8 metal drawers, 480×480×220 mm
  • Pk:
    1
Specifications
About this item
Storage organiser with 2×4 metal drawers for embedding cassettes.
  • For 2600 cassettes
  • Weight: 18 kg