Surface preparation cloth, KIMTECH PREP*
115-1554
- Packed:25/pack
- Colour:Blue
- L×W:400×400 mm
- Environmentally Preferable:
- Pk:25
This highly absorbent multipurpose cleaning and polishing cloth made from microfibre can be used wet or dry. It is effective in trapping and removing particulate matter from a variety of surfaces.
- Low linting which reduces the risk of process contamination
- Machine washable up to 300 times
- Soft and flexible
- Silicone-free, seamed edges