High Purity rNTPs, Cytiva
: Cytiva
Easy to use, time saving solutions for in vitro transcription or for reactions requiring rNTPs of high purity.
- Each lot is tested for ribonuclease contamination to ensure success in the application
- Supplied as crystalline solid or in purified water as 100 mM solution (pH 7,5)
- Available either as individual products or in a convenient set containing ATP, CTP, GTP and UTP in separate tubes
Kit contents: Adenosine 5'-Triphosphate, 100 mM Solution (ATP); Cytidine 5'-Triphosphate, 100 mM Solution (CTP); Guanosine 5'-Triphosphate, 100 mM Solution (GTP); Uridine 5'-Triphosphate, 100 mM Solution (UTP)
- Application:
- Environmentally Preferable:
Frequently Bought Together