Membrane filters
Supplier: Merck
Certificates
About this item
PVC membrane filters are specified by ASTM, NIOSH, and OSHA in air monitoring applications to analyse silica, carbon black, metals, quartz particulates and others.
- High-quality, medical-grade PVC polymer
Applications include air monitoring of particulate contaminants including silica, metals, and quartz.
Filter colour: white.
Filter surface: plain.
Specifications
- Catalog No:
- MILFPVC503700
- MILFPVC502500
- MILFPVC504700
- Type:
- PVC5
- PVC5
- PVC5
- Pore size:
- 5,00 µm
- 5,00 µm
- 5,00 µm
- Airflow rate:
- 2 l/min/cm2
- 2 l/min/cm2
- 2 l/min/cm2