ContactUsLinkComponent
Robotic OMIX™ tips for the Hamilton Microlab® Star Line
Robotic OMIX™ tips for the Hamilton Microlab® Star Line
Catalog # VARIA57403
Supplier:  VARIAN
Robotic OMIX™ tips for the Hamilton Microlab® Star Line
Catalog # VARIA57403
Supplier:  VARIAN
Specifications
  • Packed:
    5 racks, 96 tips each
  • Volume:
    300 µl
  • Tip style:
    OMIX C18
  • For use with:
    Hamilton Microlab® Star Line
  • Pk:
    5
Specifications
About this item
These tips offer excellent versatility and end-user productivity enhancements. Processing of 96 samples can be reduced to just a few minutes in certain applications. The tips have an operating volume of 300 µl, allowing flexibility in sample size.