Capillary columns, PERMABOND® Silane
Supplier: MACHEREY-NAGEL
Certificates
About this item
Special GC phase for the analysis of silanes
- Developed especially for the analysis of monomeric silanes and chlorosilanes
- Also suited for the separation of dimeric siloxanes and silazanes
- Max. temperature for isothermal operation 260 °C (0,32 mm Int.Ø) / 240 °C (0,53 mm Int.Ø)
- Max. temperature for short isotherms in a temperature program 280 °C (0,32 mm Int.Ø) / 260 °C (0,53 mm Int.Ø)
Frequently Bought Together