Cut resistant gloves, Showa 67NFW
112-5881
: Showa
- Glove size:10
- Ambidextrous / hand-specific:Hand-specific
- Disposable / reusable:Reusable
- Colour:Yellow
- Length:305 mm
- Model:67NFW
- EN norm:EN 388:2016 (3142X)
- Cuff style:Gauntlet
- Liner material:Cotton
- Pk:12 PAIR
This soft, cotton flannel liner with natural rubber coating and wrinkle finish provides superior cut, abrasion, snag, and puncture resistance.
- Wrinkle finish provides outstanding wet and dry grip
- Protects hands from chemicals and abrasion while remaining elastic and breathable
- Impermeable for working in damp or greasy environments
- Ideal replacement for leather and ordinary rubber-coated gloves
It is most suitable for mechanical and automotive applications, construction work, shipping and receiving, and wood handling.
: EN 388:2016 (3142X); Cat. II
Food contact approved
Food contact approved