Orders LinkContactUsLinkComponent
Capillary columns, CP-Lowox
Supplier:   VARIAN
Capillary columns, CP-Lowox
Supplier:  VARIAN
Certificates
Capillary columns, CP-Lowox
Capillary columns, CP-Lowox
2 Options Available Below
Product Details & Documents
CP-Lowox offers a unique solution to the chemical and petrochemical industries. It is now possible to analyze trace level oxygenate impurities in gas and liquid hydrocarbon streams.
  • Unique selectivity for a wide range of oxygenates maximises flexibility
  • No particle loss preserves detector performance
  • Suitable for process applications
This high polarity makes the column ideal for the measurement of oxygenated compounds. CP-Lowox can be used for the prevention of catalyst contamination by oxygenates, process/on-line applications or portable GC applications (ASTM D 7059).
Product Family Options
Product Information
  • Column Format
    Film thickness
    Ø int.
    Length
    Availability
    Price
  • Capillary
    10,00 µm
    0,53 mm
    10 m
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 3 948,78 €
    3 948,78 €
    1 * 1 items
       3 948,78 €
  • Capillary
    10,00 µm
    0,53 mm
    10 m