ContactUsLinkComponent
VWR® Lisbon, Visitor Spectacles
VWR® Lisbon, Visitor Spectacles
  111-0930
undefined
VWR® Lisbon, Visitor Spectacles
  111-0930
  • Type:
    Anti-UV (2-1.2), anti-scratch, anti-fog
  • Colour:
    Transparent
  • Lenses:
    Clear PC
  • Pk:
    1
Visitor glasses with wrap-around, clear polycarbonate lens and indirect ventilation.
  • Anti-scratch/anti-fog coating
  • Integrated brow guard
  • Vented side shields
Weight: 46 g
Weight: 46 g
 : Compliant with EN 166, EN 170; VWR® EN 166 FT CE (frame); VWR® 2C-1,2 1 FT (lens).