ContactUsLinkComponent
VWR® Vienna, Safety Spectacles
VWR® Vienna, Safety Spectacles
Catalog # 111-0931
Supplier:  VWR Collection
undefined
VWR® Vienna, Safety Spectacles
Catalog # 111-0931
Supplier:  VWR Collection
Specifications
  • Type:
    Anti-UV (2-1.2), anti-scratch, anti-fog
  • Colour:
    Transparent
  • Lenses:
    Clear PC
  • Pk:
    1
Specifications
About this item
Lightweight safety glasses with clear PC lens.
  • Anti-scratch/anti-fog coating
  • Pantoscopic adjustment for angle modification
  • Moulded brow guard
  • Adjustable temple length
Weight: 26 g
EN 166, EN 170; 1 FT
Certifications: Compliant with EN 166, EN 170; VWR® EN 166 FT CE (frame); VWR® 2C-1,2 1 FT (lens).