SPE columns, CHROMABOND® PSA
Supplier: MACHEREY-NAGEL
Certificates
About this item
CHROMABOND® PSA is a propylsulfonic acid modified silica cation exchanger for SPE.
- Base material: Silica
- Very strong cation exchanger
- Contrary to SCX phase (e.g. CHROMABOND® SA) no π-π interactions
Recommended for weak cations like cationic alkaloids.
Cation exchange capacity: ~0,7 meq/g
Specifications
- Catalog No:
- MANA730464
- MANA730462
- pH range:
- 2 - 8
- 2 - 8
- Surface area:
- 500 m²/g
- 500 m²/g