Haematocrit sealing material
Catalog # HECH40566010
Supplier: GLASWARENFABRIK KARL HECHT
Specifications
- Description:Hematocrit sealing wax, non-drying, in plates of polystyrene with numbered positions, for sealing hematocrit tubes
- Pk:1
Specifications
About this item
This sealing material prevents the draining of blood samples. For sealing several hundred micro hematocrit capillaries.