Distillation Glassware Sets with Ground Glass Joints 14/23, Quickfit®
Catalog # BIBB29BU/M
Supplier: SciLabware
Specifications
- Description:Organic chemistry kit, complete set (starter kit) comprising 5 items of glassware
- Environmentally Preferable:
- Pk:1
Specifications
About this item
PYREX®,Borosilicate glass.
- Comprises 5 glassware items all with size 14/23 Quickfit® ground-glass joints - giving maximum flexibility
- Scale based on starting material quantities up to 30 g
- Both reflux and distillation assemblies can be constructed
- Housed in a robust cardboard box with foam rubber liner for product protection
Starter kit - ideal for introducing students to principal techniques of preparative organic chemistry.