Distillation Glassware Sets with Ground Glass Joints 14/23, Quickfit®
Catalog # BIBB27BU
Supplier: SciLabware
Specifications
- Description:Organic chemistry kit, complete set comprising 9 items of glassware
- Environmentally Preferable:
- Pk:1
Specifications
About this item
PYREX®, Borosilicate glass.
- Comprises 9 basic items of Quickfit® glassware all with size 14/23 ground-glass joints for compatibility
- Allows the construction of at least 8 assemblies including steam distillation, reflux with addition and preparation
- Scale based on starting material quantities up to 30 g
- Housed in a robust cardboard box with foam rubber liner to provide safe storage and transit
- Supplied with a CD-ROM which includes practical details of some typical assemblies
The most popular Quickfit® set for schools, colleges and universities.