Orders LinkContactUsLinkComponent
Test tubes, Durham
Test tubes, Durham
40 mm, 8 mm
Catalog # GERB8130
Supplier:   GERBER FUNKE
Test tubes, Durham
Catalog # GERB8130
Supplier:  GERBER FUNKE
Test tubes, Durham
40 mm, 8 mm
Price
12,80 €
Log in for availability
1 * 100 items
Product Details & Documents
Glass, clear.
  • Excellent chemical resistance
  • Good temperature resistance
Documents
Specifications
  • Ext.Ø:
    8 mm
  • Length:
    40 mm