ContactUsLinkComponent
qScript™ XLT One-Step RT-qPCR ToughMix®
 :  Quantabio
undefined
qScript™ XLT One-Step RT-qPCR ToughMix®
 :  Quantabio
  • Application:
    RT-qPCR probe
  • Inhibitor-rich sample:
    Yes
  • Multiplexing:
    Yes
  • One step:
    Yes
Frequently Bought Together