Reusable earplugs, 1261/1271, Comets®
Catalog # 111-1163
Supplier: Moldex
Specifications
- Colour:Light green
- Description:Earplugs, with cord, in pocket case
- Size:U*
- Environmentally Preferable:
- Pk:50 PAIR
Specifications
About this item
These reusable earplugs provide protection against lower noise levels. They are designed to keep the user in touch with the environment.
- Air bubble in the tip and soft material provide a comfortable seal
- Plugs made of soft durable material – Washable and hygienic
- Come with handy PocketPak storage container
SNR: 25 dB
Certifications: EN 352-2:2002
Caution: * One size fits all.