SPE columns, CHROMABOND® C18 f (fast flow)
Supplier: MACHEREY-NAGEL
Certificates
About this item
CHROMABOND® C18 f is an octadecyl modified silica for a fast flow application in SPE.
- Nonpolar
- Hydrophobic and secondary interactions with polar groups of the analytes
- Suitable for nonpolar and slight polar substances
- Fast flow characteristic for viscous matrices like waste water
Recommended for nonpolar compounds with polar groups like pesticides, PCBs, pharmaceuticals and food ingredients from viscous matrices.
Specifications
- Catalog No:
- MANA730403
- MANA730009
- 554-6595
- MANA730402
- Carbon load:
- 14%
- 14%
- 14%
- 14%
- Endcapped:
- No, No
- No, No
- No, No
- No, No
- pH range:
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- Surface area:
- 500 m²/g
- 500 m²/g
- 500 m²/g
- 500 m²/g