SPE columns, CHROMABOND® C8
Supplier: MACHEREY-NAGEL
Certificates
About this item
CHROMABOND® C8 is an octyl modified silica phase for SPE.
- Slightly more polar than C18 phase
- Hydrophobic and more pronounced secondary interactions with polar compounds
- Suitable for slight nonpolar and polar substances
Recommended for polar pesticides and PCBs.
Specifications
- Catalog No:
- MANA730022
- MANA730021
- MANA738021.100M
- MANA730024G
- MANA730024
- MANA731808
- MANA730134
- Carbon load:
- 8%
- 8%
- 8%
- 8%
- 8%
- 8%
- Endcapped:
- No
- No
- No
- No
- No
- No
- pH range:
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- Surface area:
- 500 m²/g
- 500 m²/g
- 500 m²/g
- 500 m²/g
- 500 m²/g
- 500 m²/g