SPE columns, CHROMABOND® Easy
Supplier: MACHEREY-NAGEL
Certificates
About this item
CHROMABOND® Easy is a polar, bifunctionally modified polystyrene-divinylbenzene copolymer for SPE.
- Applicable for a wide range of analytes without preconditioning
- More hydrophilic than conventional polystyrene-divinylbenzene polymers
- Easily wettable with water
Applications: Polar pesticides (acidic, neutral, basic) or phenols from water; drug analysis from urine, blood, serum or from tablets and creams.
Specifications
- Catalog No:
- MANA730751
- MANA730757
- MANA730756
- MANA730754.250
- MANA730759
- MANA730753
- MANA730755
- MANA730754
- MANA730755.250
- pH range:
- 1 - 14
- 1 - 14
- 1 - 14
- 1 - 14
- 1 - 14
- 1 - 14
- 1 - 14
- 1 - 14
- 1 - 14
- Surface area:
- 650 - 700 m²/g
- 650 - 700 m²/g
- 650 - 700 m²/g
- 650 - 700 m²/g
- 650 - 700 m²/g
- 650 - 700 m²/g
- 650 - 700 m²/g
- 650 - 700 m²/g
- 650 - 700 m²/g