Orders LinkContactUsLinkComponent
Homogenisers, behrotest®
Homogenisers, behrotest®
Supplier:   BEHR
Homogenisers, behrotest®
Supplier:  BEHR
Certificates
Homogenisers, behrotest®
2 Options Available Below
Product Details & Documents
Homogenisers for pretreatment, sample splitting and homogenisation of heterogeneous water samples in accordance with German waste water quality control standards.
  • The sample vessel is protected against damage by the homogeniser frame, which has two handles for easy carrying
  • Sample vessel volumes from 5 to 25 litre permit multiple sample collection (mixed samples)
  • Graduated for easy measurement of sample volume
  • Adjustable-speed magnetic stirrer
  • Withdrawl of desired volume of homogenized sample by means of a PTFE stopcock
  • Can be operated from a 12 V auto battery using an optional voltage converter
Product Family Options
Product Information
  • Availability
    Price
  • 331-3310
    Homogeniser, 15 litre capacity, QMR 15
    Homogeniser, 15 litre capacity, QMR 15
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 3 672,12 €
    3 672,12 €
    1 * 1 items
       3 672,12 €
  • BEHRB00218333
    Homogeniser, 20 litre capacity, QMR 20
    Homogeniser, 20 litre capacity, QMR 20
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 4 134,31 €
    4 134,31 €
    1 * 1 items
       4 134,31 €
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • Behrotest glass vessel, 15 l
    Behrotest glass vessel, 15 l
    331-3312
    Log in for availability
    1 * 1 items - 1 162,29 €