Cation exchange HPLC columns, Bio MAb
Supplier: Agilent
Certificates
About this item
These HPLC columns feature a unique resin specifically designed for providing higher resolution with smaller particles and charge based separations of monoclonal antibodies.
- A packing support composed of a rigid, spherical, highly cross-linked polystyrene divinylbenzene (PS/DVB) non-porous bead
- Particles grafted with a hydrophilic, polymeric layer, virtually eliminating non-specific binding of antibody proteins
- Particles use a different process to layer the weak cation exchange phase to the particle making it a higher density
- Its thorough characterisation of monoclonal antibodies includes the identification and monitoring of acidic and basic isoforms
Specifications
- Catalog No:
- HEWL5190-2406
- HEWL5190-2405
- HEWL5190-2403
- HEWL5190-2419
- HEWL5190-2413
- HEWL5190-2415
- HEWL5190-2412
- HEWL5190-2411
- HEWL5190-2407
- HEWL5190-2420
- HEWL5190-2414
- pH range:
- 2 - 12
- 2 - 12
- 2 - 12
- 2 - 12
- 2 - 12
- 2 - 12
- 2 - 12
- 2 - 12
- 2 - 12
- 2 - 12
- 2 - 12
- Temperature limit:
- 80 ºC
- 80 ºC
- 80 ºC
- 80 ºC
- 80 ºC
- 80 ºC
- 80 ºC
- 80 ºC
- 80 ºC
- 80 ºC
- 80 ºC